Kalium(3-(methylthio)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3215-1G TB-3215-25G TB-3215-5G
COBBTB-3215-1GEA 74.2 EUR
COBBTB-3215-1G COBBTB-3215-25G COBBTB-3215-5G
Kalium(3-(methylthio)phenyl)trifluoroborat
Potassium-3-(methylthio)phenyltrifluoroborate
Formel: C₇H₇BF₃KS
Molecular Weight: 230,1 g/mol
MDL: MFCD04115747
CAS-Nummer: 850623-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR