Kalium(4-(ethylthio)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3247-1G TB-3247-5G TB-3247-25G
COBBTB-3247-1GEA 38.2 EUR
COBBTB-3247-1G COBBTB-3247-5G COBBTB-3247-25G
Kalium(4-(ethylthio)phenyl)trifluoroborat
Potassium-4-(ethylthio)phenyltrifluoroborate
Formel: C₈H₉BF₃KS
Molecular Weight: 244,12 g/mol
MDL: MFCD04115771
CAS-Nummer: 850623-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR