Kalium(2-(methylsulfonyl)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Potassium trifluoro(2-(methylsulphonyl)phenyl)borate, Potassium (2-methylsulphonylphenyl)trifluoroborate

TB-3233-1G TB-3233-5G
COBBTB-3233-1GEA 124 EUR
COBBTB-3233-1G COBBTB-3233-5G
Kalium(2-(methylsulfonyl)phenyl)trifluoroborat
Potassium-2-(methylsulphonyl)phenyltrifluoroborate
Formel: C₇H₇BF₃KO₂S
Molecular Weight: 262,1000 g/mol
MDL: MFCD04115760
CAS-Nummer: 850623-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR