Kalium(4-(methylsulfonyl)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3203-1G TB-3203-5G
COBBTB-3203-1GEA 0 EUR
COBBTB-3203-1G COBBTB-3203-5G
Kalium(4-(methylsulfonyl)phenyl)trifluoroborat
Potassium-4-(methylsulphonyl)phenyltrifluoroborate
Formel: C₇H₇BF₃KO₂S
MDL: MFCD04115738
CAS-Nummer: 850623-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR