Methyl-4-chlor-3,5-dibromsalicylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-chloro-3,5-dibromo-2-hydroxybenzoate

CA-4399-5G CA-4399-1G
COBBCA-4399-5GEA 149 EUR
COBBCA-4399-5G COBBCA-4399-1G
Methyl-4-chlor-3,5-dibromsalicylat
Methyl-4-chlor-3,5-dibromsalicylat
Formel: C₈H₅Br₂ClO₃
MDL: MFCD09475886
CAS-Nummer: 941294-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR