3-Brom-5-nitrobenzamid
Lieferant: COMBI-BLOCKS
CA-4348-5G
CA-4348-1G
COBBCA-4348-5GEA
230
EUR
COBBCA-4348-5G
COBBCA-4348-1G
3-Brom-5-nitrobenzamid
3-Brom-5-nitrobenzamid
Formel:
C₇H₅BrN₂O₃ |
MDL:
MFCD01529620 CAS-Nummer: 54321-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...