3-Brom-4-(trifluormethoxy)benzonitril

Lieferant: COMBI-BLOCKS

CA-4774-5G CA-4774-1G CA-4774-250M
COBBCA-4774-5GEA 1350 EUR
COBBCA-4774-5G COBBCA-4774-1G COBBCA-4774-250M
3-Brom-4-(trifluormethoxy)benzonitril
3-Brom-4-(trifluormethoxy)benzonitril
Formel: C₈H₃BrF₃NO
MDL: MFCD13195683
CAS-Nummer: 191602-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR