2-Brom-5-fluorbenzamid

Lieferant: COMBI-BLOCKS

CA-4432-1G CA-4432-5G
COBBCA-4432-1GEA 204 EUR
COBBCA-4432-1G COBBCA-4432-5G
2-Brom-5-fluorbenzamid
2-Brom-5-fluorbenzamid
Formel: C₇H₅BrFNO
Molecular Weight: 218.02 g/mol
MDL: MFCD03094322
CAS-Nummer: 1006-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR