3-Brom-4-fluorbenzamid

Lieferant: COMBI-BLOCKS

CA-4614-5G CA-4614-1G
COBBCA-4614-5GEA 166 EUR
COBBCA-4614-5G COBBCA-4614-1G
3-Brom-4-fluorbenzamid
3-Brom-4-fluorbenzamid
Formel: C₇H₅BrFNO
Molecular Weight: 218,0200 g/mol
MDL: MFCD03094431
CAS-Nummer: 455-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR