3-Brom-N-cyclohexyl-4-fluorbenzamid
Lieferant: COMBI-BLOCKS
CA-4607-5G
CA-4607-25G
CA-4607-1G
COBBCA-4607-5GEA
303
EUR
COBBCA-4607-5G
COBBCA-4607-25G
COBBCA-4607-1G
3-Brom-N-cyclohexyl-4-fluorbenzamid
3-Brom-N-cyclohexyl-4-fluorbenzamid
Formel:
C₁₃H₁₅BrFNO |
MDL:
MFCD11504867 CAS-Nummer: 1065074-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...