2-Brom-N-(2,4-difluorphenyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6087-5G HC-6087-1G
COBBHC-6087-5GEA 0 EUR
COBBHC-6087-5G COBBHC-6087-1G
2-Brom-N-(2,4-difluorphenyl)benzolsulfonamid
2-Brom-N-(2,4-difluorphenyl)benzolsulfonamid
Formel: C₁₂H₈BrF₂NO₂S
MDL: MFCD09475878
CAS-Nummer: 941294-30-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR