5-Brom-2-iodbenzonitril

Lieferant: COMBI-BLOCKS

OS-7683-25G OS-7683-5G OS-7683-1G
COBBOS-7683-25GEA 308 EUR
COBBOS-7683-25G COBBOS-7683-5G COBBOS-7683-1G
5-Brom-2-iodbenzonitril
5-Brom-2-iodbenzonitril
Formel: C₇H₃BrIN
Molecular Weight: 307,92 g/mol
MDL: MFCD00672950
CAS-Nummer: 121554-10-7
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR