2-Brom-N-(2,4-dimethylphenyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6085-1G HC-6085-5G
COBBHC-6085-1GEA 98.3 EUR
COBBHC-6085-1G COBBHC-6085-5G
2-Brom-N-(2,4-dimethylphenyl)benzolsulfonamid
2-Brom-N-(2,4-dimethylphenyl)benzolsulfonamid
Formel: C₁₄H₁₄BrNO₂S
MDL: MFCD05617812
CAS-Nummer: 941294-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR