Methyl-2-formylbenzoat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 2-formylbenzoate

10X-0992.1 10X-0992.10 10X-0992.5 10X-0992.500
BIOT10X-0992.1EA 0 EUR
BIOT10X-0992.1 BIOT10X-0992.10 BIOT10X-0992.5 BIOT10X-0992.500
Methyl-2-formylbenzoat
Methyl-2-formylbenzoat
Formel: C₉H₈O₃
Molecular Weight: 164.1580 g/mol
MDL: MFCD00204231
CAS-Nummer: 4122-56-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR