4-(Diethylamino)salicylaldehyd 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-(Diethylamino)-2-hydroxybenzaldehyde
209480050
209480250
ACRO209480050EA
22
EUR
ACRO209480050
ACRO209480250
4-(Diethylamino)salicylaldehyd 98%
4-(Diethylamino)salicylaldehyd
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193,25 g/mol Schmelzpunkt: 60…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003326 CAS-Nummer: 17754-90-4 EINECS: 241-745-3 |
Spezifikation Testergebnisse
Appearance | Pink to light brown to purple Powder |
Infrared spectrum | Conforms |
Melting point | 60 to 64 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...