4-(Diethylamino)salicylaldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-(Diethylamino)-2-hydroxybenzaldehyde
M-Clarity™ Qualitätsstufe = MQ200
225681-25G
225681-5G
SIAL225681-25GEA
65.4
EUR
SIAL225681-25G
SIAL225681-5G
4-(Diethylamino)salicylaldehyd, Sigma-Aldrich®
4-(Diethylamino)salicylaldehyd
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193,25 g/mol |
MDL:
MFCD00003326 CAS-Nummer: 17754-90-4 EINECS: 241-745-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...