2-(Trimethylsilyloxy)benzaldehyd
Lieferant: CHEMPUR SILANE
SILS19899.10
SILS19899.25
SILS19899.100
SILNSILS19899.10EA
0
EUR
SILNSILS19899.10
SILNSILS19899.25
SILNSILS19899.100
2-(Trimethylsilyloxy)benzaldehyd
2-(Trimethylsilyloxy)benzaldehyd
Formel:
C₁₀H₁₄O₂Si Molecular Weight: 194.31 g/mol |
MDL:
MFCD00040721 CAS-Nummer: 1078-31-5 EINECS: 214-081-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...