4-(Trimethylsilyloxy)benzaldehyd

Lieferant: CHEMPUR SILANE

SILS19900.10 SILS19900.25 SILS19900.100
SILNSILS19900.10EA 0 EUR
SILNSILS19900.10 SILNSILS19900.25 SILNSILS19900.100
4-(Trimethylsilyloxy)benzaldehyd
4-(Trimethylsilyloxy)benzaldehyd
Formel: C₁₀H₁₄O₂Si
MDL: MFCD00040723
CAS-Nummer: 1012-12-0
EINECS: 213-789-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR