3,5-Dichlor-4-isopropoxyphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3,5-Dichloro-4-isopropoxyphenylboronic acid, pinacol ester
PN-6231-5G
PN-6231-1G
COBBPN-6231-5GEA
884
EUR
COBBPN-6231-5G
COBBPN-6231-1G
3,5-Dichlor-4-isopropoxyphenylboronsäurepinakolester
3,5-Dichlor-4-isopropoxyphenylboronsäurepinakolester
Formel:
C₁₅H₂₁BCl₂O₃ |
MDL:
MFCD15143609 CAS-Nummer: 1218790-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...