2-Chlor-6-fluor-3-nitrobenzonitril

Lieferant: CHEMPUR

CD07793.250 CD07793.1
CHMPCD07793.250EA 0 EUR
CHMPCD07793.250 MAYBCD07793.1
2-Chlor-6-fluor-3-nitrobenzonitril
2-Chlor-6-fluor-3-nitrobenzonitril
Formel: C₇H₂ClFN₂O₂
MDL: MFCD00053834
CAS-Nummer: 226419-18-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR