3-Chlor-4-fluorphenylhydrazin Hydrochlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(3-Chloro-4-fluorophenyl)hydrazine hydrochloride
A10175.06
A10175.14
A10175.06EA
61.9
EUR
A10175.06
A10175.14
3-Chlor-4-fluorphenylhydrazin Hydrochlorid ≥98%
3-Chlor-4-fluorphenylhydrazin Hydrochlorid
Purity: 98%
CAS No.: 175135-74-7
Molecular Formula: C6H62FN2.HCl
Formula Weight: 197.04
Melting Point: ca 210° dec.
MDL No.: MFCD00052267
Formel:
C₆H₇Cl₂FN₂ Molecular Weight: 197.04 g/mol Schmelzpunkt: ∼ 210 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052267 CAS-Nummer: 175135-74-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...