3-Chlor-4-fluorphenylhydrazin Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: (3-Chloro-4-fluorophenyl)hydrazine hydrochloride

008468-1G 008468-5G
FLUO008468-1GEA 0 EUR
FLUO008468-1G FLUO008468-5G
3-Chlor-4-fluorphenylhydrazin Hydrochlorid
3-Chlor-4-fluorphenylhydrazin Hydrochlorid
Formel: C₆H₇Cl₂FN₂
Molecular Weight: 197.04 g/mol
MDL: MFCD00052267
CAS-Nummer: 175135-74-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR