4-Cyanphenylhydrazin Hydrochlorid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Hydrazinylbenzonitrile hydrochloride
B22157.06
B22157.14
B22157.22
B22157.06EA
43
EUR
B22157.06
B22157.14
B22157.22
4-Cyanphenylhydrazin Hydrochlorid ≥97%
4-Cyanphenylhydrazin Hydrochlorid
Purity: 97%
CAS No.: 2863-98-1
Molecular Formula: C7H7N3.HCl
Formula Weight: 169.61
Melting Point: ca 243° dec.
Beilstein Reference: 3565486
Formel:
C₇H₈ClN₃ Molecular Weight: 169.61 g/mol Schmelzpunkt: ∼ 243 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00673994 CAS-Nummer: 2863-98-1 |