4,5-Dimethoxy-2-nitrobenzonitril

Lieferant: CHEMPUR

FL13146.POR 013146-25G 013146-5G
CHMPFL13146.POREA 0 EUR
CHMPFL13146.POR FLUO013146-25G FLUO013146-5G
4,5-Dimethoxy-2-nitrobenzonitril
4,5-Dimethoxy-2-nitrobenzonitril
Formel: C₉H₈N₂O₄
MDL: MFCD00017564
CAS-Nummer: 102714-71-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR