Methyl-4-aminosalicylat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 4-amino-2-hydroxybenzoate

11P-706.500 11P-706.1 11P-706.10 11P-706.5
BIOT11P-706.500EA 0 EUR
BIOT11P-706.500 BIOT11P-706.1 BIOT11P-706.10 BIOT11P-706.5
Methyl-4-aminosalicylat
Methyl-4-aminosalicylat
Formel: C₈H₉NO₃
Molecular Weight: 167.18 g/mol
MDL: MFCD00088091
CAS-Nummer: 4136-97-4
EINECS: 223-958-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR