Methyl-4-amino-3-hydroxybenzoat 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 4-amino-3-hydroxybenzoate

L05687.09 L05687.04
L05687.09EA 285 EUR
L05687.09 L05687.04
Methyl-4-amino-3-hydroxybenzoat 98%
Methyl-4-amino-3-hydroxybenzoat

Purity: 98% CAS No.: 63435-16-5 Molecular Formula: C8H9NO3 Formula Weight: 167.17 Melting Point: 118-122° Beilstein Reference: 608716

Formel: C₈H₉NO₃
Molecular Weight: 167,17 g/mol
Schmelzpunkt: 118…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017093
CAS-Nummer: 63435-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR