Methyl-5-methoxysalicylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-hydroxy-5-methoxybenzoate

456683-25ML 456683-5ML
SIAL456683-25MLEA 300 EUR
SIAL456683-25ML SIAL456683-5ML
Methyl-5-methoxysalicylat, Sigma-Aldrich®
Methyl-5-methoxysalicylat
Formel: C₉H₁₀O₄
Molecular Weight: 182,1733 g/mol
MDL: MFCD00017185
CAS-Nummer: 2905-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR