Kalium(3-fluor-4-methoxyphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Potassium trifluoro(3-fluoro-4-methoxyphenyl)borate, Potassium(3-Fluoro-4-methoxyphenyl)trifluoroborate

TB-3229-1G TB-3229-5G
COBBTB-3229-1GEA 49.7 EUR
COBBTB-3229-1G COBBTB-3229-5G
Kalium(3-fluor-4-methoxyphenyl)trifluoroborat
Potassium-3-fluoro-4-methoxyphenyltrifluoroborate
Formel: C₇H₆BF₄KO
Molecular Weight: 232,0300 g/mol
MDL: MFCD04115757
CAS-Nummer: 850623-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR