2-(Trifluormethoxy)benzoesäurehydrazid

Lieferant: FLUOROCHEM

Synonyme: 2-(Trifluoromethoxy)benzhydrazide

007542-1G 007542-5G
FLUO007542-1GEA 55.4 EUR
FLUO007542-1G FLUO007542-5G
2-(Trifluormethoxy)benzoesäurehydrazid
2-(Trifluoromethoxy)benzoic acid hydrazide
Formel: C₈H₇F₃N₂O₂
MDL: MFCD00204180
CAS-Nummer: 175277-19-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR