4-(Trifluormethoxy)benzoesäurehydrazid
Lieferant: FLUOROCHEM
Synonyme:
4-(Trifluoromethoxy)benzohydrazide, 4-(Trifluoromethoxy)benzhydrazide
CFF687.5
007543-1G
007543-5G
CHMPCFF687.5EA
401
EUR
CHMPCFF687.5
FLUO007543-1G
FLUO007543-5G
4-(Trifluormethoxy)benzoesäurehydrazid
4-(Trifluoromethoxy)benzoic acid hydrazide
Formel:
C₈H₇F₃N₂O₂ Molecular Weight: 220.15 g/mol |
MDL:
MFCD00204234 CAS-Nummer: 175277-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...