5-(Trifluormethoxy)salicylsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Hydroxy-5-(trifluoromethoxy)benzoic acid

013025-1G 013025-5G
FLUO013025-1GEA 174 EUR
FLUO013025-1G FLUO013025-5G
5-(Trifluormethoxy)salicylsäure
5-(Trifluormethoxy)salicylsäure
Formel: C₈H₅F₃O₄
MDL: MFCD01091003
CAS-Nummer: 129644-57-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR