4-(Trifluormethoxy)phenylhydrazin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: (4-(Trifluoromethoxy)phenyl)hydrazine hydrochloride

CFF1280. FL17116.POR 017116-1G 017116-25G 017116-5G
CHMPCFF1280.EA 0 EUR
CHMPCFF1280. CHMPFL17116.POR FLUO017116-1G FLUO017116-25G FLUO017116-5G
4-(Trifluormethoxy)phenylhydrazin Hydrochlorid
4-(Trifluormethoxy)phenylhydrazin Hydrochlorid
Formel: C₇H₈ClF₃N₂O
Molecular Weight: 228.60 g/mol
MDL: MFCD00053033
CAS-Nummer: 133115-72-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR