3-Ethoxy-2-fluor-6-nitroanisol
Lieferant: COMBI-BLOCKS
AN-1381-1G
AN-1381-5G
COBBAN-1381-1GEA
0
EUR
COBBAN-1381-1G
COBBAN-1381-5G
3-Ethoxy-2-fluor-6-nitroanisol
3-Ethoxy-2-fluor-6-nitroanisol
Formel:
C₉H₁₀FNO₄ |
MDL:
MFCD11504833 CAS-Nummer: 1072945-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...