Methyl-5-iodsalicylat ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 2-hydroxy-5-iodobenzoate

B22048.09 B22048.18
B22048.09EA 43.5 EUR
B22048.09 B22048.18
Methyl-5-iodsalicylat ≥99%
Methyl-5-iodsalicylat

Purity: 99% CAS No.: 4068-75-1 Molecular Formula: C8H7IO3 Formula Weight: 278.05 Melting Point: 75-78° MDL No.: MFCD00016462

Formel: C₈H₇IO₃
Molecular Weight: 278,05 g/mol
Schmelzpunkt: 75…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016462
CAS-Nummer: 4068-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR