Lithium-3,5-diiodsalicylat, Supelco®

Lieferant: Merck

D3635-25G D3635-5G
D3635-25GEA 507 EUR
D3635-25G D3635-5G
Lithium-3,5-diiodsalicylat, Supelco®
Lithium-3,5-diiodsalicylat
Formel: C₇H₃I₂LiO₃
MDL: MFCD00002443
CAS-Nummer: 653-14-5
EINECS: 211-496-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c4e0323603de9e37f8b156e40d732f1f38f6721&vid=7f236d58f9aff4974b303dc6dacd673c9745c6de&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c9cd6b548e0e2c08b5542d8f560ea55a17c5d41&vid=257f32a649b2b5c6830b8e3c072bd615304250d7&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR