5-Amino-2,4,6-triiodisophthalsäure 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 5-Amino-2,4,6-triiodoisophthalic Acid

B22178.09 B22178.18 B22178.30
B22178.09EA 26.2 EUR
B22178.09 B22178.18 B22178.30
5-Amino-2,4,6-triiodisophthalsäure 95%
5-Amino-2,4,6-triiodisophthalsäure

Purity: 95% CAS No.: 35453-19-1 Molecular Formula: C8H4I3NO4 Formula Weight: 558.84 Melting Point: 265-270° MDL No.: MFCD00190167

Formel: C₈H₄I₃NO₄
Molecular Weight: 558.84 g/mol
Schmelzpunkt: 265…270 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00190167
CAS-Nummer: 35453-19-1
EINECS: 252-575-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR