4-Nitrophenylsulfurpentafluorid

Lieferant: FLUOROCHEM

CFF1245. 008100-1G 008100-5G
CHMPCFF1245.EA 0 EUR
CHMPCFF1245. FLUO008100-1G FLUO008100-5G
4-Nitrophenylsulfurpentafluorid
4-Nitrophenylsulfurpentafluorid
Formel: C₆H₄F₅NO₂S
Molecular Weight: 249.16 g/mol
MDL: MFCD00221619
CAS-Nummer: 2613-27-6
EINECS: 447-610-7
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR