Cyclopropylphenylsulfid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Cyclopropyl(phenyl)sulphane

L12321.03 L12321.06
L12321.03EA 26.3 EUR
L12321.03 L12321.06
Cyclopropylphenylsulfid ≥98%
Cyclopropylphenylsulfid

Purity: 98+% CAS No.: 14633-54-6 Molecular Formula: C9H10S Formula Weight: 150.24 Flash Point: 91°(195°F) Refractive Index: 1.5810 Odour: Stench Boiling Point: 61-62°/1mm Beilstein Reference: 2075701 UN No.: 3334 MDL No.: MFCD00009684

Formel: C₉H₁₀S
Molecular Weight: 150,24 g/mol
Siedepunkt: 61…62 °C (1 mmHg)
Dichte: 1,045
Flammpunkt: 91 °C (195 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009684
CAS-Nummer: 14633-54-6
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR