Cyclopropylphenylsulfid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cyclopropyl(phenyl)sulphane

M-Clarity™ Qualitätsstufe = MQ100

227242-25G 227242-5G
SIAL227242-25GEA 268 EUR
SIAL227242-25G SIAL227242-5G
Cyclopropylphenylsulfid, Sigma-Aldrich®
Cyclopropylphenylsulfid
Formel: C₉H₁₀S
Molecular Weight: 150,24 g/mol
MDL: MFCD00009684
CAS-Nummer: 14633-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR