2-Amino-4-(4-bromphenyl)thiazol

Lieferant: FLUOROCHEM

011076-25G 011076-5G
FLUO011076-25GEA 195 EUR
FLUO011076-25G FLUO011076-5G
2-Amino-4-(4-bromphenyl)thiazol
2-Amino-4-(4-bromophenyl)thiazole
Formel: C₉H₇BrN₂S
Molecular Weight: 255.1343 g/mol
MDL: MFCD00051953
CAS-Nummer: 2103-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR