2-Amino-4-(4-chlorphenyl)thiazol 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-(4-Chlorophenyl)thiazol-2-amine

L14067.03 L14067.06
L14067.03EA 25.7 EUR
L14067.03 L14067.06
2-Amino-4-(4-chlorphenyl)thiazol 98%
2-Amino-4-(4-chlorophenyl)thiazole

Purity: 98% CAS No.: 2103-99-3 Molecular Formula: C9H7ClN2S Formula Weight: 210.69 Melting Point: 168-170° Beilstein Reference: 153889

Formel: C₉H₇ClN₂S
Molecular Weight: 210.69 g/mol
Schmelzpunkt: 168…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047058
CAS-Nummer: 2103-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR