2-(4-Methylphenyl)-4-thiazolcarbonsäure
Lieferant: BIONET RESEARCH
Synonyme:
2-(p-Tolyl)thiazole-4-carboxylic acid
2G-402S.500
2G-402S.1
2G-402S.5
2G-402S.10
BIOT2G-402S.500EA
0
EUR
BIOT2G-402S.500
BIOT2G-402S.1
BIOT2G-402S.5
BIOT2G-402S.10
2-(4-Methylphenyl)-4-thiazolcarbonsäure
2-(4-Methylphenyl)-4-thiazolecarboxylic acid
Formel:
C₁₁H₉NO₂S Molecular Weight: 219.26 g/mol |
MDL:
MFCD01936002 CAS-Nummer: 17228-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...