2-(4-Methoxyphenyl)-4-thiazolcarbonsäure

Lieferant: BIONET RESEARCH

1W-0286.500 1W-0286.1 1W-0286.5 1W-0286.10
BIOT1W-0286.500EA 0 EUR
BIOT1W-0286.500 BIOT1W-0286.1 BIOT1W-0286.5 BIOT1W-0286.10
2-(4-Methoxyphenyl)-4-thiazolcarbonsäure
2-(4-Methoxyphenyl)-4-thiazolecarboxylic acid
Formel: C₁₁H₉NO₃S
MDL: MFCD03791177
CAS-Nummer: 57677-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR