2-Mercapto-4-phenylthiazol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Phenylthiazole-2-thiol

L05829.06 L05829.14
L05829.06EA 78 EUR
L05829.06 L05829.14
2-Mercapto-4-phenylthiazol ≥98%
2-Mercapto-4-phenylthiazole

Purity: 98% CAS No.: 2103-88-0 Molecular Formula: C9H7NS2 Formula Weight: 193.30 Odour: Odourless Melting Point: 172-175° Beilstein Reference: 128288 MDL No.: MFCD00014508

Formel: C₉H₇NS₂
Molecular Weight: 193,30 g/mol
Schmelzpunkt: 172…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014508
CAS-Nummer: 2103-88-0
EINECS: 218-274-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR