2-[(Trimethylsilyl)ethinyl]benzaldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-((Trimethylsilyl)ethynyl)benzaldehyde
M-Clarity™ Qualitätsstufe = MQ100
590614-25G
590614-5G
SIAL590614-25GEA
354
EUR
SIAL590614-25G
SIAL590614-5G
2-[(Trimethylsilyl)ethinyl]benzaldehyd, Sigma-Aldrich®
2-[(Trimethylsilyl)ethinyl]benzaldehyd
Formel:
C₁₂H₁₄OSi Molecular Weight: 202,324 g/mol |
MDL:
MFCD04039984 CAS-Nummer: 77123-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...