4-Methoxycarbonyl-2-nitrophenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 3-nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
PN-2461-5G
PN-2461-25G
PN-2461-1G
COBBPN-2461-5GEA
404
EUR
COBBPN-2461-5G
COBBPN-2461-25G
COBBPN-2461-1G
4-Methoxycarbonyl-2-nitrophenylboronsäurepinakolester
4-Methoxycarbonyl-2-nitrophenylboronsäurepinakolester
Formel:
C₁₄H₁₈BNO₆ Molecular Weight: 307,1100 g/mol |
MDL:
MFCD09878423 CAS-Nummer: 957065-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...