4-(Hydroxymethyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Hydroxymethyl)benzolboronsäure(tetramethylethylen)ester

M-Clarity™ Qualitätsstufe = MQ100

702927-1G 702927-5G
SIAL702927-1GEA 52 EUR
SIAL702927-1G SIAL702927-5G
4-(Hydroxymethyl)phenylboronsäurepinakolester, Sigma-Aldrich®
4-(Hydroxymethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₉BO₃
Molecular Weight: 234,10 g/mol
MDL: MFCD09837617
CAS-Nummer: 302348-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR