4-Methyl-3-nitrophenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4-Methyl-3-nitrophenyl)boronic acid, 4-Methyl-3-nitrobenzeneboronic acid

L17052.06 L17052.03
L17052.06EA 82 EUR
L17052.06 L17052.03
4-Methyl-3-nitrophenylboronsäure ≥98%
4-Methyl-3-nitrophenylboronsäure

Purity: 98% CAS No.: 80500-27-2 Molecular Formula: C7H8BNO4 Formula Weight: 180.96 Melting Point: ca 250° Beilstein Reference: 3279018

Formel: C₇H₈BNO₄
Molecular Weight: 180.96 g/mol
Schmelzpunkt: ∼ 250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191550
CAS-Nummer: 80500-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR