4-Methyl-3-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Methyl-3-nitrophenyl)boronic acid, 4-Methyl-3-nitrobenzeneboronic acid

BB-2334-25G BB-2334-5G BB-2334-1G
COBBBB-2334-25GEA 293 EUR
COBBBB-2334-25G COBBBB-2334-5G COBBBB-2334-1G
4-Methyl-3-nitrophenylboronsäure
4-Methyl-3-nitrophenylboronsäure
Formel: C₇H₈BNO₄
Molecular Weight: 180.96 g/mol
MDL: MFCD00191550
CAS-Nummer: 80500-27-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR