4-(Hydroxymethyl)phenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-(Hydroxymethyl)benzeneboronic acid
M-Clarity™ Qualitätsstufe = MQ100
512338-10G
512338-1G
SIAL512338-10GEA
248
EUR
SIAL512338-10G
SIAL512338-1G
4-(Hydroxymethyl)phenylboronsäure, Sigma-Aldrich®
4-(Hydroxymethyl)phenylboronsäure
Formel:
C₇H₉BO₃ Molecular Weight: 151,96 g/mol |
MDL:
MFCD00792672 CAS-Nummer: 59016-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...